[(Carboxymethoxy)amino](oxo)acetic acid

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: N-(Carboxymethoxy)oxamic acid

OR40325-100MG OR40325-1G OR40325-500MG
77409-518EA 259.84 USD
77409-518 77409-520 77409-522
[(Carboxymethoxy)amino](oxo)acetic acid
[(Carboxymethoxy)amino](oxo)acetic acid
Formula: C₄H₅NO₆
MW: 163.086 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17168268
CAS Number: 177902-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR