1-(Tributylstannyl)-2,4-dimethylbenzene

Supplier: APOLLO SCIENTIFIC

Synonyms: tributyl-(2,4-dimethylphenyl)stannane

OR399041-1G OR399041-250MG
77409-416EA 303.6 USD
77409-416 77409-418
1-(Tributylstannyl)-2,4-dimethylbenzene
1-(Tributylstannyl)-2,4-dimethylbenzene
Formula: C₂₀H₃₆Sn
Storage Temperature: Ambient
CAS Number: 1301268-27-8
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR