1-(Tributylstannyl)-3,5-bistrifluoromethylbenzene

Supplier: APOLLO SCIENTIFIC

Synonyms: [3,5-bis(trifluoromethyl)phenyl]-tributyl-stannane

OR399038-1G OR399038-250MG
77409-404EA 333.16 USD
77409-404 77409-406
1-(Tributylstannyl)-3,5-bistrifluoromethylbenzene
1-(Tributylstannyl)-3,5-bistrifluoromethylbenzene
Formula: C₂₀H₃₀F₆Sn
Storage Temperature: Ambient
CAS Number: 717918-62-2
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR