4-(Tributylstannyl)-1,2-methylenedioxybenzene

Supplier: APOLLO SCIENTIFIC

Synonyms: 1,3-benzodioxol-5-yl(tributyl)stannane

OR399036-1G OR399036-250MG
77409-396EA 303.6 USD
77409-396 77409-398
4-(Tributylstannyl)-1,2-methylenedioxybenzene
4-(Tributylstannyl)-1,2-methylenedioxybenzene
Formula: C₁₉H₃₂O₂Sn
Storage Temperature: Ambient
CAS Number: 124010-49-7
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR