4-(Tributylstannyl)-1-ethylbenzene

Supplier: APOLLO SCIENTIFIC

Synonyms: tributyl-(4-ethylphenyl)stannane

OR399032-1G OR399032-250MG
77409-388EA 303.6 USD
77409-388 77409-390
4-(Tributylstannyl)-1-ethylbenzene
4-(Tributylstannyl)-1-ethylbenzene
Formula: C₂₀H₃₆Sn
Storage Temperature: Ambient
CAS Number: 198224-53-2
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR