(2-Cyanophenyl)methanesulfonyl chloride
Supplier: APOLLO SCIENTIFIC
OR322865-1G
OR322865-250MG
77409-186EA
275.08
USD
77409-186
77409-188
(2-Cyanophenyl)methanesulfonyl chloride
(2-Cyanophenyl)methanesulfonyl chloride
Formula:
C₈H₆ClNO₂S Storage Temperature: Ambient |
MDL Number:
MFCD07355846 CAS Number: 51045-34-2 UN: 2923 ADR: 8+6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...