7-Bromo-2,3-dihydro-1H-isoindol-1-one 95%

Supplier: APOLLO SCIENTIFIC

OR305302-250MG OR305302-1G
77408-894EA 23.44 USD
77408-894 77408-892
7-Bromo-2,3-dihydro-1H-isoindol-1-one 95%
7-Bromo-2,3-dihydro-1H-isoindol-1-one
Formula: C₈H₆BrNO
Storage Temperature: Ambient
MDL Number: MFCD11848600
CAS Number: 200049-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR