1-(5-Bromo-3-methylpyridin-2-yl)ethanone

Supplier: APOLLO SCIENTIFIC

OR300405-1G OR300405-250MG
77408-656EA 678.08 USD
77408-656 77408-658
1-(5-Bromo-3-methylpyridin-2-yl)ethanone
1-(5-Bromo-3-methylpyridin-2-yl)ethanone
Formula: C₈H₈BrNO
MW: 214.059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18261721
CAS Number: 1211533-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR