5-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazolo[3,4-b]pyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR300398-1G OR300398-250MG
77408-640EA 326.02 USD
77408-640 77408-642
5-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazolo[3,4-b]pyridine
5-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazolo[3,4-b]pyridine
Formula: C₁₁H₁₂BrN₃O
MW: 282.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22689759
CAS Number: 1256957-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR