(5-Bromo-3-chloropyridin-2-yl)methanamine hydrochloride 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR300395-1G OR300395-250MG OR300395-100MG
77408-634EA 467.64 USD
77408-634 77408-636 77408-632
(5-Bromo-3-chloropyridin-2-yl)methanamine hydrochloride 95%
(5-Bromo-3-chloropyridin-2-yl)methanamine hydrochloride
Formula: C₆H₆BrClN₂·ClH
MW: 257.943 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22689891
CAS Number: 1416714-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR