(1-(Tetrahydro-2H-pyran-2-yl)-1H-indazol-5-yl)boronic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR300384-1G OR300384-250MG
77408-624EA 183.38 USD
77408-624 77408-626
(1-(Tetrahydro-2H-pyran-2-yl)-1H-indazol-5-yl)boronic acid
(1-(Tetrahydro-2H-pyran-2-yl)-1H-indazol-5-yl)boronic acid
Formula: C₁₂H₁₅BN₂O₃
MW: 246.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922966
CAS Number: 1562245-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR