3-Bromo-4-chloro-1H-pyrrolo[3,2-c]pyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR300374-1G OR300374-250MG
77408-606EA 311.76 USD
77408-606 77408-608
3-Bromo-4-chloro-1H-pyrrolo[3,2-c]pyridine
3-Bromo-4-chloro-1H-pyrrolo[3,2-c]pyridine
Formula: C₇H₄BrClN₂
MW: 231.477 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11007847
CAS Number: 947238-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR