Ethyl 2-(4-ethylphenyl)thiazole-4-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR300148-1G OR300148-250MG
77408-576EA 506.4 USD
77408-576 77408-578
Ethyl 2-(4-ethylphenyl)thiazole-4-carboxylate
Ethyl 2-(4-ethylphenyl)thiazole-4-carboxylate
Formula: C₁₄H₁₅NO₂S
MW: 261.339 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06738347
CAS Number: 885278-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR