5-Hydroxy-3-nitrobenzo[b]furan 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Nitro-1-benzofuran-5-ol, 3-Nitrobenzo[b]furan-5-ol, 5-Hydroxy-3-nitro-1-benzofuran, 3-Nitrobenzofuran-5-ol

OR29193-1G OR29193-500MG
77408-364EA 88.74 USD
77408-364 77408-366 77408-368
5-Hydroxy-3-nitrobenzo[b]furan 97%
5-Hydroxy-3-nitrobenzo[b]furan
Formula: C₈H₅NO₄
MW: 179.13 g/mol
Melting Pt: 164…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00114881
CAS Number: 126318-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR