2-Methyl-4-(piperazin-1-yl)quinoline

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-(2-Methylquinolin-4-yl)piperazine

OR27760-1G OR27760-250MG
77407-982EA 186.44 USD
77407-982 77407-984
2-Methyl-4-(piperazin-1-yl)quinoline
2-Methyl-4-(piperazin-1-yl)quinoline
Formula: C₁₄H₁₇N₃
MW: 227.305 g/mol
Melting Pt: 100…101 °C
Storage Temperature: Ambient
MDL Number: MFCD01935745
CAS Number: 82241-22-3
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR