6-(Dimethylamino)pyridine-3-boronic acid hydrate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Borono-2-(dimethylamino)pyridine hydrate

OR2767-1G OR2767-500MG
77407-960EA 65.36 USD
77407-960 77407-962
6-(Dimethylamino)pyridine-3-boronic acid hydrate
6-(Dimethylamino)pyridine-3-boronic acid hydrate
Formula: C₇H₁₁BN₂O₂·H₂O
MW: 184.001 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06656059
CAS Number: 1256355-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR