6-Methylimidazo[2,1-b][1,3]thiazole-5-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR25897-1G OR25897-5G
77407-686EA 124.3 USD
77407-686 77407-688
6-Methylimidazo[2,1-b][1,3]thiazole-5-carboxylic acid
6-Methylimidazo[2,1-b][1,3]thiazole-5-carboxylic acid
Formula: C₇H₆N₂O₂S
MW: 182.2 g/mol
Melting Pt: 164…165 °C
Storage Temperature: Ambient
MDL Number: MFCD02083132
CAS Number: 77628-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR