Ethyl 2-(2,4-dichlorophenyl)-4-hydroxythiazole-5-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR2426-1G OR2426-250MG OR2426-50MG
77407-456EA 359.64 USD
77407-456 77407-458 77407-462
Ethyl 2-(2,4-dichlorophenyl)-4-hydroxythiazole-5-carboxylate
Ethyl 2-(2,4-dichlorophenyl)-4-hydroxythiazole-5-carboxylate
Formula: C₁₂H₉Cl₂NO₃S
MW: 318.176 g/mol
Melting Pt: 150…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00816792
CAS Number: 262856-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR