2-(4-Morpholino)benzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Morpholinobenzoic acid

OR23383-1G OR23383-25G OR23383-5G
77407-296EA 82.54 USD
77407-296 77407-298 77407-300
2-(4-Morpholino)benzoic acid
2-(4-Morpholino)benzoic acid
Formula: C₁₁H₁₃NO₃
Melting Pt: 151…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00527976
CAS Number: 42106-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR