Ethyl 4-(chlorosulfonyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate

Supplier: APOLLO SCIENTIFIC
Danger

OR23364-100MG OR23364-250MG
77407-282EA 57 USD
77407-282 77407-284 77407-286
Ethyl 4-(chlorosulfonyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate
Ethyl 4-(chlorosulfonyl)-3,5-dimethyl-1H-pyrrole-2-carboxylate
Formula: C₉H₁₂ClNO₄S
Melting Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD03407326
CAS Number: 368869-88-9
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR