1-[3-(Bromomethyl)phenyl]-1H-pyrrole

Supplier: APOLLO SCIENTIFIC
Danger

OR23350-1G OR23350-250MG
77407-262EA 203.76 USD
77407-262 77407-264
1-[3-(Bromomethyl)phenyl]-1H-pyrrole
1-[3-(Bromomethyl)phenyl]-1H-pyrrole
Formula: C₁₁H₁₀BrN
Melting Pt: 47…50 °C
Storage Temperature: Ambient
MDL Number: MFCD03086141
CAS Number: 112596-36-8
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR