Bicyclo[2.2.1]heptane-1-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR22825-100MG OR22825-250MG OR22825-500MG
77406-956EA 225.16 USD
77406-956 77406-958 77406-960
Bicyclo[2.2.1]heptane-1-carboxylic acid
Bicyclo[2.2.1]heptane-1-carboxylic acid
Formula: C₈H₁₂O₂
Storage Temperature: Freezer
MDL Number: MFCD00180984
CAS Number: 18720-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR