3-(1H-Imidazol-1-yl)benzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(3-Formylphenyl)-1H-imidazole, 3-(1-Imidazolyl)benzaldehyde

OR2268-1G OR2268-25G OR2268-5G
77406-934EA 172.18 USD
77406-934 77406-936 77406-938
3-(1H-Imidazol-1-yl)benzaldehyde
3-(1H-Imidazol-1-yl)benzaldehyde
Formula: C₁₀H₈N₂O
MW: 172.183 g/mol
Melting Pt: 80…81 °C
Storage Temperature: Ambient
MDL Number: MFCD09836178
CAS Number: 127404-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR