3-Amino-4-hydroxy-N,N-dimethylbenzenesulfonamide 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR22180-100MG OR22180-1G
77406-826EA 21.26 USD
77406-826 77406-828
3-Amino-4-hydroxy-N,N-dimethylbenzenesulfonamide 95%
3-Amino-4-hydroxy-N,N-dimethylbenzenesulfonamide
Formula: C₈H₁₂N₂O₃S
MW: 216.257 g/mol
Melting Pt: 156…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00179913
CAS Number: 24962-75-2
EINECS: 246-548-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR