Diphenyl(vinyl)phosphine oxide

Supplier: APOLLO SCIENTIFIC
Warning

OR22135-1G OR22135-5G OR22135-10G
77406-794EA 103.92 USD
77406-794 77406-796 77406-792
Diphenyl(vinyl)phosphine oxide
Diphenyl(vinyl)phosphine oxide
Formula: C₁₄H₁₃OP
MW: 228.226 g/mol
Melting Pt: 115…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00114913
CAS Number: 2096-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR