6-Phenylnicotinonitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR2090-100MG OR2090-1G OR2090-250MG
77406-576EA 28.52 USD
77406-576 77406-578 77406-580
6-Phenylnicotinonitrile 97%
6-Phenylnicotinonitrile
Formula: C₁₂H₈N₂
Melting Pt: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD06637420
CAS Number: 39065-54-8
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR