2-(3-Methyl-1,2,4-oxadiazol-5-yl)benzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-(2-Carboxyphenyl)-3-methyl-1,2,4-oxadiazole

OR2076-1G OR2076-250MG
77406-570EA 260.82 USD
77406-570 77406-572
2-(3-Methyl-1,2,4-oxadiazol-5-yl)benzoic acid
2-(3-Methyl-1,2,4-oxadiazol-5-yl)benzoic acid
Formula: C₁₀H₈N₂O₃
MW: 204.182 g/mol
Melting Pt: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD08741426
CAS Number: 475105-77-2
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR