3-(Piperazin-1-yl)benzoic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(3-Carboxyphenyl)piperazine

OR1886-1G OR1886-5G
77406-422EA 206.82 USD
77406-422 77406-424
3-(Piperazin-1-yl)benzoic acid 97%
3-(Piperazin-1-yl)benzoic acid
Formula: C₁₁H₁₄N₂O₂
MW: 206.241 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03444416
CAS Number: 446831-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR