2-Chloro-3,5-dimethylpyridine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Chloro-3,5-lutidine

OR18609-100MG OR18609-250MG
77406-362EA 32.6 USD
77406-362 77406-364
2-Chloro-3,5-dimethylpyridine
2-Chloro-3,5-dimethylpyridine
Formula: C₇H₈ClN
MW: 141.598 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12400986
CAS Number: 72093-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR