3-Bromo-4-chlorobenzeneacetic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Bromo-4-chlorophenylacetic acid, 2-(3-Bromo-4-chlorophenyl)acetic acid

OR17683-1G OR17683-25G OR17683-5G
77406-088EA 21.32 USD
77406-088 77406-090 77406-092
3-Bromo-4-chlorobenzeneacetic acid 95%
3-Bromo-4-chlorobenzeneacetic acid
Formula: C₈H₆BrClO₂
MW: 249.489 g/mol
Melting Pt: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD18390582
CAS Number: 90004-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR