N-(2-Hydroxyethyl)methacrylamide 96%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
HEMAm, N-(2-Hydroxyethyl)-2-methylprop-2-enamide
OR17009-1G
OR17009-25G
OR17009-5G
77405-904EA
47.88
USD
77405-904
77405-906
77405-908
N-(2-Hydroxyethyl)methacrylamide 96%
N-(2-Hydroxyethyl)methacrylamide
Formula:
C₆H₁₁NO₂ MW: 129.157 g/mol Density: 0.95 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00080529 CAS Number: 5238-56-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...