1-(7-Nitro-1,1,3,3,6-pentamethylindan-5-yl)-1H-pyrrole

Supplier: APOLLO SCIENTIFIC

Synonyms: 1-(2,3-Dihydro-7-nitro-1,1,3,3,6-pentamethyl-1H-inden-5-yl)-1H-pyrrole

OR16908-1G OR16908-5G
77405-846EA 166.06 USD
77405-846 77405-848
1-(7-Nitro-1,1,3,3,6-pentamethylindan-5-yl)-1H-pyrrole
1-(7-Nitro-1,1,3,3,6-pentamethylindan-5-yl)-1H-pyrrole
Formula: C₁₈H₂₂N₂O₂
MW: 298.379 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00206198
CAS Number: 657405-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR