3-Methyl-2-nitrobenzaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Nitro-m-tolualdehyde, 3-Formyl-2-nitrotoluene

OR16689-1G OR16689-250MG OR16689-5G
77405-816EA 187.46 USD
77405-816 77405-818 77405-820
3-Methyl-2-nitrobenzaldehyde 97%
3-Methyl-2-nitrobenzaldehyde
Formula: C₈H₇NO₃
MW: 165.146 g/mol
Melting Pt: 59…61 °C
Storage Temperature: Ambient
MDL Number: MFCD08703364
CAS Number: 5858-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR