(4-Oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl)acetic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR16618-250MG OR16618-1G
77405-774EA 156.9 USD
77405-774 77405-772
(4-Oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl)acetic acid
(4-Oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl)acetic acid
Formula: C₁₁H₁₁NO₃S
MW: 237.275 g/mol
Melting Pt: 225…230 °C
Storage Temperature: Ambient
MDL Number: MFCD01416244
CAS Number: 17547-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR