4-(Chloromethyl)-1H-imidazole hydrochloride 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR15569-1G OR15569-250MG OR15569-5G
77405-460EA 117.16 USD
77405-460 77405-462 77405-464
4-(Chloromethyl)-1H-imidazole hydrochloride 95%
4-(Chloromethyl)-1H-imidazole hydrochloride
Formula: C₄H₅ClN₂·ClH
MW: 153.01 g/mol
Melting Pt: 139…145 °C
Storage Temperature: Ambient
MDL Number: MFCD01721661
CAS Number: 38585-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR