1-[(Thien-2-yl)methyl]piperidine-4-carboxaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Formyl-1-[(thien-2-yl)methyl]piperidine, 2-[(4-Formylpiperidin-1-yl)methyl]thiophene

OR14936-1G OR14936-250MG
77405-192EA 309.6 USD
77405-192 77405-194
1-[(Thien-2-yl)methyl]piperidine-4-carboxaldehyde 97%
1-[(Thien-2-yl)methyl]piperidine-4-carboxaldehyde
Formula: C₁₁H₁₅NOS
MW: 209.308 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09879953
CAS Number: 926921-80-4
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR