3-Methylfur-2-yl isocyanate 90%

Supplier: APOLLO SCIENTIFIC
Danger

OR14918-100MG OR14918-1G OR14918-500MG
77405-180EA 344.36 USD
77405-180 77405-182 77405-184
3-Methylfur-2-yl isocyanate 90%
3-Methylfur-2-yl isocyanate

May form small amount of sediment.

Formula: C₆H₅NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09879906
CAS Number: 921938-67-2
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR