2-Bromo-1,5-dimethyl-1H-imidazole 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR1343-100MG
OR1343-1G
OR1343-5G
77404-640EA
21.4
USD
77404-640
77404-642
77404-644
2-Bromo-1,5-dimethyl-1H-imidazole 97%
2-Bromo-1,5-dimethyl-1H-imidazole
Formula:
C₅H₇BrN₂ MW: 175.026 g/mol Melting Pt: 40…42 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06659902 CAS Number: 235426-31-0 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...