2-Phenyl-1-(piperazin-1-yl)ethan-1-one

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: [2-Oxo-2-(piperazin-1-yl)ethyl]benzene, 1-(Phenylacetyl)piperazine

OR13308-100MG OR13308-250MG
77404-618EA 135.2 USD
77404-618 77404-620 77404-622
2-Phenyl-1-(piperazin-1-yl)ethan-1-one
2-Phenyl-1-(piperazin-1-yl)ethan-1-one
Formula: C₁₂H₁₆N₂O
MW: 204.268 g/mol
Boiling Pt: 190…191 °C (17 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01566912
CAS Number: 88372-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR