4-(4-Methoxybenzyl)piperidine hydrochloride 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-[(Piperidin-4-yl)methyl]anisole hydrochloride

OR12871-250MG OR12871-5G OR12871-1G
77404-454EA 98.82 USD
77404-454 77404-456 77404-452
4-(4-Methoxybenzyl)piperidine hydrochloride 95%
4-(4-Methoxybenzyl)piperidine hydrochloride
Formula: C₁₃H₁₉NO·ClH
MW: 241.757 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03840137
CAS Number: 37581-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR