2-(Morpholin-4-yl)-1-phenylethylamine 95+%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-(2-Amino-2-phenylethyl)morpholine, α-[(Morpholin-4-yl)methyl]benzylamine

OR12713-100MG OR12713-1G OR12713-250MG
77404-410EA 99.84 USD
77404-410 77404-412 77404-414
2-(Morpholin-4-yl)-1-phenylethylamine 95+%
2-(Morpholin-4-yl)-1-phenylethylamine
Formula: C₁₂H₁₈N₂O
MW: 206.284 g/mol
Boiling Pt: 130…134 °C (2 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01862532
CAS Number: 38060-08-1
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR