3-(4-Chlorophenoxy)benzenesulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Chloro-3'-(chlorosulphonyl)diphenyl ether, 3-(4-Chlorophenoxy)benzene-1-sulphonyl chloride

OR12560-100MG OR12560-1G OR12560-250MG
77404-368EA 209.18 USD
77404-368 77404-370 77404-372
3-(4-Chlorophenoxy)benzenesulfonyl chloride
3-(4-Chlorophenoxy)benzenesulfonyl chloride
Formula: C₁₂H₈Cl₂O₃S
Storage Temperature: Ambient
MDL Number: MFCD02089485
CAS Number: 501697-55-8
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR