Benzyl 2-acetamido-2-deoxy-α-D-glucopyranoside

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Benzyl 2-(acetylamino)-2-deoxy-α-D-glucopyranoside, N-((2S,3R,4R,5S,6R)-2-(Benzyloxy)-4,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)acetamide

OR1254T-100MG OR1254T-1G OR1254T-250MG
77404-358EA 21.26 USD
77404-358 77404-360 77404-362
Benzyl 2-acetamido-2-deoxy-α-D-glucopyranoside
Benzyl 2-acetamido-2-deoxy-α-D-glucopyranoside
Formula: C₁₅H₂₁NO₆
MW: 311.33 g/mol
Melting Pt: 179…182 °C
Storage Temperature: Freezer
MDL Number: MFCD00070371
CAS Number: 13343-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR