2-(2-Methylphenyl)pyrrolidine 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR12510-1G OR12510-250MG OR12510-5G
77404-334EA 316.86 USD
77404-334 77404-336 77404-338
2-(2-Methylphenyl)pyrrolidine 95%
2-(2-Methylphenyl)pyrrolidine
Formula: C₁₁H₁₅N
Storage Temperature: Refrigerator
MDL Number: MFCD02663451
CAS Number: 129540-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR