3-(1-Methyl-1H-pyrazol-5-yl)aniline 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR12382-1G OR12382-250MG
77404-288EA 467.64 USD
77404-288 77404-290
3-(1-Methyl-1H-pyrazol-5-yl)aniline 95%
3-(1-Methyl-1H-pyrazol-5-yl)aniline
Formula: C₁₀H₁₁N₃
Melting Pt: 79…80 °C
Storage Temperature: Ambient
MDL Number: MFCD09702400
CAS Number: 910037-08-0
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR