5-(Aminomethyl)benzo[b]furan 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 5-(Aminomethyl)-1-benzofuran, (Benzo[b]furan-5-yl)methylamine

OR12314-1G OR12314-250MG
77404-264EA 327.06 USD
77404-264 77404-266
5-(Aminomethyl)benzo[b]furan 97%
5-(Aminomethyl)benzo[b]furan
Formula: C₉H₉NO
Melting Pt: 32…34 °C
Storage Temperature: Ambient
MDL Number: MFCD04974039
CAS Number: 37798-08-6
UN: 3259
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR