Methyl 5-bromo-4-methylthiophene-2-carboxylate 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR11283-1G OR11283-250MG OR11283-5G
77403-874EA 85.58 USD
77403-874 77403-876 77403-878
Methyl 5-bromo-4-methylthiophene-2-carboxylate 98%
Methyl 5-bromo-4-methylthiophene-2-carboxylate
Formula: C₇H₇BrO₂S
Melting Pt: 39…40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04038959
CAS Number: 54796-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR