2-(2-Chloro-3,4-dimethoxyphenyl)acetic acid

Supplier: APOLLO SCIENTIFIC

OR111438-250MG OR111438-1G
77403-774EA 89.66 USD
77403-774 77403-772
2-(2-Chloro-3,4-dimethoxyphenyl)acetic acid
2-(2-Chloro-3,4-dimethoxyphenyl)acetic acid
Formula: C₁₀H₁₁ClO₄
MW: 230.645 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21606658
CAS Number: 6834-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR