3,4-Dichlorobenzyl iodide

Supplier: APOLLO SCIENTIFIC
Danger

OR111348-1G OR111348-5G
77403-728EA 119 USD
77403-728 77403-730
3,4-Dichlorobenzyl iodide
3,4-Dichlorobenzyl iodide
Formula: C₇H₅Cl₂I
MW: 286.925 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410705
CAS Number: 142523-67-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR