4,6-Dichloro-2-iodopyrimidine

Supplier: APOLLO SCIENTIFIC
Warning

OR111089-1G OR111089-250MG
77403-638EA 323.98 USD
77403-638 77403-640
4,6-Dichloro-2-iodopyrimidine
4,6-Dichloro-2-iodopyrimidine
Formula: C₄HCl₂IN₂
Storage Temperature: Ambient
MDL Number: MFCD22207084
CAS Number: 1266105-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR